Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LTBP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551930
Description
Immunogen sequence: ERSPNLRRSS AAGEGTLARA QPPAPQSPPA PQSPPAGTLS GLSQTHPSQQ HVGLSRTVRL HPTATASSQL SSNALPPGPG LEQRDGTQQA VPLEHPSSPW GLNLTEKIKK IKIVFTPTIC KQTCARGHCA NSCERG Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 62%.
The protein encoded by this gene belongs to the family of latent transforming growth factor (TGF)-beta binding proteins (LTBP), which are extracellular matrix proteins with multi-domain structure. This protein is the largest member of the LTBP family possessing unique regions and with most similarity to the fibrillins. It has thus been suggested that it may have multiple functions: as a member of the TGF-beta latent complex, as a structural component of microfibrils, and a role in cell adhesion.
Specifications
LTBP2 | |
Polyclonal | |
Unconjugated | |
LTBP2 | |
AW208642; C14orf141; GLC3D; latent transforming growth factor beta binding protein 2; latent-transforming growth factor beta-binding protein 2; LTBP2; LTBP-2; LTBP3; MSPKA; MSTP031; WMS3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4053 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14767 | |
LTBP2 | |
Recombinant protein corresponding to Human LTBP2. Recombinant protein control fragment (Product #RP-89084). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction