Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTF Mouse anti-Human, Clone: 119CT80.1.1, Abnova™

Mouse monoclonal antibody raised against human LTF.

Manufacturer:  Abnova Corporation MAB12285

Catalog No. 89-934-491


Add to cart

Description

Description

This gene is a member of the transferrin family of genes and its protein product is found in the secondary granules of neutrophils. The protein is a major iron-binding protein in milk and body secretions with an antimicrobial activity, making it an important component of the non-specific immune system. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis. [provided by RefSeq]

Specifications

Specifications

LTF
119CT80.1.1
Mouse monoclonal antibody raised against human LTF.
In PBS (0.09% sodium azide)
GIG12/HLF2/LF
Mouse
IgG1, kappa
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (1:50-1:100) Immunofluorescence (1:10-1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
LTF
Recombinant protein corresponding to human LTF.
Protein G purification
RUO
Primary
4057
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit