Tocris™ Lu AA 47070

Prodrug of a potent and selective adenosine A2A receptor antagonist

Manufacturer: Tocris™ 4783/10

 View more versions of this product

Catalog No. 47-831-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors Lu AA 47070
    Purity >98
    Molecular Weight 463.39
    Quantity 10mg
    Product Type Lu AA 47070

    Prodrug of a potent and selective adenosine A2A receptor antagonist (Ki values are 5.9, 260, 410 and <10,000nM for A2A, A2B, A1 and A3 receptors respectively. Reverses parkinsonian motor impairment and motivational effects produced by dopamine D2 receptor blockade in rats. Orally bioavailable.

    • Chemical Name: 4-[(3,3-Dimethyl-1-oxobutyl)amino]-3,5-difluoro-N-[3-[(phosphonooxy)methyl]-2(3H)-thiazolylidene]benzamide
    • Formula: C17H20F2N3O6PS
    • CAS Number: 913842-25-8
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit