Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ LU AA33810
Potent NPY Y5 receptor antagonist
$250.56 - $1033.56
Specifications
Purity | >98% |
---|---|
Product Type | LU AA33810 |
Molecular Weight (g/mol) | 423.62 |
Inhibitors | LU AA33810 |
Description
Potent neuropeptide Y (NPY) Y5 receptor antagonist (Ki = 1.5 nM in vitro). Displays ≥ 3300-fold affinity for Y5 over Y1, Y2 and Y4 receptors. Also binds human 5-HT2B and 5-HT1A receptors (Ki values are 247 and 478 nM respectively). Exerts anxiolytic- and antidepressant-like effects in rat models of stress sensitivity.Specifications
>98% | |
LU AA33810 | |
423.62 | |
LU AA33810 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title