Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lubrol MP Biomedicals

$95.16 - $159.56

Chemical Identifiers

CAS 8047-15-2
Physical Form Waxy Solid
Molecular Formula HO(CH2CH2O)n C16H33 n = ∽23
Synonym Polyethylene glycolmonocetyl ether
Chemical Name or Material Lubrol, Molecular Biology
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN19529983 MP Biomedicals, IncSupplier Diversity Partner
0219529983
250g Each for $159.56
Check Availability
Add to cart
ICN19529980 MP Biomedicals, IncSupplier Diversity Partner
0219529980
100g Each for $95.16
Check Availability
Add to cart
Specifications Description & Specifications

Chemical Identifiers

8047-15-2
HO(CH2CH2O)n C16H33 n = ∽23
Lubrol, Molecular Biology
Waxy Solid
Polyethylene glycolmonocetyl ether

Specifications

8047-15-2
HO(CH2CH2O)n C16H33 n = ∽23
Off-white
Waxy Solid
Polyethylene glycolmonocetyl ether
Lubrol, Molecular Biology

Safety and Handling

Recommended Storage : Store at room temperature (15° to 30°C)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit