Learn More
LUC7L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 170851AP
Catalog No. 50-173-1376
Add to cart
Description
Description
May bind to RNA via its Arg/Ser-rich domain.
Specifications
Specifications
LUC7L
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9CYI4, Q9NQ29
LUC7 LIKE, LUC7 like (S. cerevisiae), LUC7B1, LUC7L, LUC7L1, Putative SR protein LUC7B1, SR+89
Rabbit
IgG
150 μL
-20°C
Primary
360503, 55692, 66978
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LUC7L
Liquid
Luc7l
LUC7L Fusion Protein Ag9170
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat