Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LUC7L3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553816
Description
Immunogen sequence: LAEVERRIRR GHARLALSQN QQSSGAAGPT GKNEEKIQVL TDKIDVLLQQ IEELGSEGKV EEAQGMMKLV EQLKEERELL RSTTSTIESF A Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Binds cAMP regulatory element DNA sequence. May play a role in RNA splicing.
Specifications
LUC7L3 | |
Polyclonal | |
Unconjugated | |
LUC7L3 | |
3300001P08Rik; cAMP regulatory element-associated protein 1; cisplatin resistance associated overexpressed protein; cisplatin resistance-associated overexpresse protein-like protein; cisplatin resistance-associated overexpressed protein; cisplatin resistance-associated-overexpressed protein; CRA; CREAP1; CREAP-1; CRE-associated protein 1; Crop; FLJ11063; hLuc7A; LUC7 like 3 pre-mRNA splicing factor; Luc7A; LUC7L3; LUC7-like 3; LUC7-like 3 (S. cerevisiae); LUC7-like 3 pre-mRNA splicing factor; luc7-like protein 3; O48; OA48-18; okadaic acid-inducible phosphoprotein OA48-18; RGD1307981; zgc:136380; zgc:158165 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51747 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95232 | |
LUC7L3 | |
Recombinant protein corresponding to Human LUC7L3. Recombinant protein control fragment (Product #RP-92019). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction