Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lucifer yellow CH, Dilithium salt, ≥99%, >95%, MP Biomedicals

Catalog Number p-4495715
Click to view available options
Quantity:
10 mg
25 mg
100 mg
67769-47-5
C13H9Li2N5O9S2
457.241
RPKCZJYDUKVMGF-UHFFFAOYSA-L
6-amino-2[(Hyrazinocarbonyl)amino]-2,3-dihydro-1,3-dioxo-1H-Benz[de] Isoquinoline-5,8-disulfonic acid dilithium salt
124220108
CHEBI:52106
dilithium;6-amino-2-(hydrazinecarbonylamino)-1,3-dioxobenzo[de]isoquinoline-5,8-disulfonate
[Li+].[Li+].C1=C(C=C2C3=C1C(=O)N(C(=O)C3=CC(=C2N)S(=O)(=O)[O-])NC(=O)NN)S(=O)(=O)[O-]
67769-47-5
C13H9Li2N5O9S2
457.241
RPKCZJYDUKVMGF-UHFFFAOYSA-L
6-amino-2[(Hyrazinocarbonyl)amino]-2,3-dihydro-1,3-dioxo-1H-Benz[de] Isoquinoline-5,8-disulfonic acid dilithium salt
124220108
CHEBI:52106
dilithium;6-amino-2-(hydrazinecarbonylamino)-1,3-dioxobenzo[de]isoquinoline-5,8-disulfonate
[Li+].[Li+].C1=C(C=C2C3=C1C(=O)N(C(=O)C3=CC(=C2N)S(=O)(=O)[O-])NC(=O)NN)S(=O)(=O)[O-]
Assay Percent Range >95%
Formula Weight 457.2
Percent Purity ≥99%
Color Orange to Yellow
Quantity 10 mg
Physical Form Powder
Chemical Name or Material Lucifer yellow CH, Dilithium salt, For HPLC analysis
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit