Learn More
BioVision IncSupplier Diversity Partner Luciferase Reporter Assay Kit: Luciferase Reporter Assay Kit: Convenient & Sensitive Assay for Detecting Luciferase Activity in transfected eukaryotic cells within 20-25 min. 200 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K801200
Firefly luciferase has been used as a sensitive reporter for studying gene regulation and function. Luciferase provides a 1000-fold increase in sensitivity in comparison to the standard chloramphenicol acetyltransferase (CAT) assay. Firefly luciferase catalyzes the oxidative carboxylation of luciferin, a reaction wz the highest efficiency of any known bioluminescence reaction. The light emission from the reaction can be recorded using a luminometer. BioVision's Luciferase Reporter Assay Kit provides a simple means for detecting luciferase activity in transfected eukaryotic cells. For measurement of expressed luciferase in vitro, luciferase is first extracted from transfected cells through cell lysis. Then CoA, ATP, Mg2+ and buffer are added to the lysate (Inclusion of CoA yields a nearly constant light emission rather than typical flash Kinetics). The luminescent reaction is then triggered by an injection of luciferin, & the emitted light is recorded by a luminometer
Catalog No. NC0176173