Learn More
Ludwig Schneider™ Exact-Temp Bottle-Encased Nonmercury Incubator Thermometers
Certified chamber thermometers, graduated in 0.1°C or 0.5°C
Manufacturer:  *LUDWIG SCHNEIDER I31001S
 View more versions of this product
Catalog No. 15-060-275
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
The items indicated contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Specifications Description & Specifications
Specifications
Total
190mm
+25° to +45°C
7.48 in.
0.1°C