Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LUF 6283, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
92933-48-7
C8H12N2O2
168.196
ZJTXSGLJNBAMJS-UHFFFAOYSA-N
5-butyl-2h-pyrazole-3-carboxylic acid,3-butyl-1h-pyrazole-5-carboxylic acid,3-butylpyrazole-5-carboxylic acid,5-n-butyl-1h-pyrazole-3-carboxylic acid,d0ff9f,5-butyl-1h-pyrazol-3-carboxylic acid,1h-pyrazole-3-carboxylic acid, 5-butyl,luf hplc
6485181
5-butyl-1H-pyrazole-3-carboxylic acid
CCCCC1=CC(=NN1)C(=O)O
This item is not returnable. View return policy
92933-48-7
C8H12N2O2
168.196
ZJTXSGLJNBAMJS-UHFFFAOYSA-N
5-butyl-2h-pyrazole-3-carboxylic acid,3-butyl-1h-pyrazole-5-carboxylic acid,3-butylpyrazole-5-carboxylic acid,5-n-butyl-1h-pyrazole-3-carboxylic acid,d0ff9f,5-butyl-1h-pyrazol-3-carboxylic acid,1h-pyrazole-3-carboxylic acid, 5-butyl,luf hplc
6485181
5-butyl-1H-pyrazole-3-carboxylic acid
CCCCC1=CC(=NN1)C(=O)O

HCA2 (GPR109A) partial agonist

Partial agonist of hydroxycarboxylic acid receptor 2 (HCA2) (Ki = 0.55 μM). Thought to lower plasma very-low-density lipoprotein (VLDL)-triglyceride levels by affecting hepatic VLDL production in normolipidemic C57BL/6 mice.
Quantity 50mg
Formula Weight 168.19
Percent Purity >98%
Chemical Name or Material LUF 6283

Recommended Storage : Store at +4°C