Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORS Nasco Lufkin™ 700 Series Power Tape

Powerful dual magnets serve as a pick-up tool

Manufacturer:  ORS Nasco 182L725MAG

Catalog No. 19-313-619


Add to cart

Description

Description

  • Magnetic end hook is a time-saver for framers, HVAC installers, and more
  • Non-slip co-molded rubber grip
  • Non-slip rubber blade lock button
  • Blade W x L: 1 in. x 25 ft. (2.5cm x 7.6m)

Specifications

Specifications

Power Tape
0.8 lb.
ABS Plastic, Magnetic, Rubber
Tape
Yellow
0.36 kg
1 Ea.
1 in.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit