Learn More
Abnova™ LUM 293T Cell Transient Overexpression Lysate (Denatured) (T03)
Human overexpression lysate
Manufacturer:  Abnova™ H00004060T03
Catalog No. 89-951-259
Add to cart
Description
Description
Specifications
Specifications
293T
NM_002345
pCMV-LUM full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LUM 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 37.29kDa
Transient overexpression cell lysate was tested with anti-LUM antibody (Mfr. No. H00004060-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human