Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Luminaris Color HiGreen qPCR Master Mix, Fluorescein

Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.

$216.50 - $2500.00

Specifications

Detection Method Detection Method: SYBR, PCR Method: qPCR
For Use With (Equipment) BioRad MyiQ, BioRad iCycler iQ, BioRad iQ5
Sample Type DNA, dsDNA
For Use With (Application) Real time PCR (qPCR), gene expression analysis, siRNA validation, genotyping and pathogen detection
Storage Requirements Note that SYBR Green and ROX dyes are light sensitive. Exposure to light should be minimized.
View More Specs

Products 4
Catalog Number Mfr. No. No. of Tests Price Quantity & Availability  
Catalog Number Mfr. No. No. of Tests Price Quantity & Availability  
FERK0381 View Documents Thermo Scientific™
K0381
250 Reactions Each for $216.50
Add to cart
 
FERK0383 View Documents Thermo Scientific™
K0383
1250 Reactions Each for $921.50
Add to cart
 
FERK0384 View Documents Thermo Scientific™
K0384
5000 Reactions Each for $2,500.00
Add to cart
 
FERK0382 View Documents Thermo Scientific™
K0382
500 Reactions Each for $391.00
Add to cart
 
Description

Description

  • Blue master mix and Yellow Sample Buffer for easy pipetting
  • UDG in the master mix to prevent carry-over contamination
  • Equal performance of blue and colorless master mixes
  • Versions with premixed ROX or fluorescein for different real-time platforms
  • Standard cycling protocols
  • Excellent qPCR performance
  • Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity: single copy detection
  • Wide linear range: accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience: ready-to-use 2X master mix
Specifications

Specifications

Detection Method: SYBR, PCR Method: qPCR
DNA, dsDNA
Note that SYBR Green and ROX dyes are light sensitive. Exposure to light should be minimized.
BioRad MyiQ, BioRad iCycler iQ, BioRad iQ5
Real time PCR (qPCR), gene expression analysis, siRNA validation, genotyping and pathogen detection

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit