Learn More
American Educational Products Lung/Breath Volume Kits Products for Science Education
Investigate pulmonary fitness.
Manufacturer:  American Educational Products 6050
 View more versions of this product
Catalog No. S16205
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Description
Description
  • Lung Volume Kit allows you to measure lung volume with reusable components
  • Breath Volume Kit measures tidal and respiratory minute volume
Specifications
Specifications
To measure lung volume with reusable components
Lung Volume Kit
3
Durable plastic bag calibrated to 6l, mouthpiece and mouthpiece holder
9
Education