Learn More
Lung Specific Antigen Mouse anti-Human, Alexa Fluor™ 750, Clone: LSA120, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP234677AF750
Catalog No. NP234677A70
Add to cart
Description
Description
Lung Specific Antigen Monoclonal antibody specifically detects Lung Specific Antigen in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen).
Specifications
Specifications
Lung Specific Antigen
LSA120
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
Protein extract of human lung cells
0.1 mL
Primary
LSA120 MAb is directed against a lung cell antigen. It shows a well-defined reactivity with cells of the lung lineage at all stages of maturation. Non-lung lineages are negative by immuno-histology. Although the exact identity of its antigen has yet to be determined, it has been shown to be distinct from MOC-52 and UM-A9 proteins. This MAb is useful in the identification of normal and neoplastic human lung cells.