Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lung Tissue Slides, Adult Normal, Female - age 83

Tissue was fixed in formalin immediately after excision and embedded in paraffin. 

Manufacturer:  Novus Biologicals™ NBP242696

Catalog No. NBP242696


Add to cart

Specifications

Specifications

Lung
From female, aged 83
5 Slides
Normal
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C. Do not freeze.

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit