Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lutetium foil, 0.127mm (0.005 in.) thick, 99.9% (REO)

Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
7439-94-3
Lu
174.967
MFCD00011098
OHSVLFRHMCKCQY-UHFFFAOYSA-N
lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces
23929
CHEBI:33382
lutetium
[Lu]
7439-94-3
Lu
174.967
MFCD00011098
OHSVLFRHMCKCQY-UHFFFAOYSA-N
lutecium,unii-5h0doz21uj,5h0doz21uj,foil,foil, 0.127mm 0.005in thick reo,cassiopium,lutecio,cassiopeium,atom,pieces
23929
CHEBI:33382
lutetium
[Lu]
  • Packaged under argon
Quantity 25 x 25 mm
Weight ≈0.78g/25x25mm
UN Number UN3178
Odor Odorless
Packaging Mylar under Argon
Physical Form Foil
Percent Purity 99.9%
Color Silver to Gray
Chemical Name or Material Lutetium foil, 0.127mm (0.005 in.) thick
Assay (REO)

GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-103-0

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures