Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lutetium(III) chloride, 99.9%, (trace metal basis), anhydrous, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
10099-66-8
Cl3Lu
MFCD00011099
10099-66-8
Cl3Lu
MFCD00011099
For ultra trace analysis
Assay Percent Range 99.9%
Linear Formula LuCl3
Formula Weight 281.33
Physical Form Powder
Grade Trace Metal Basis
Packaging Glass bottle
Total Trace Metal Impurities 0.15% max.
Color White to Gray
Quantity 5g
Chemical Name or Material Lutetium(III) chloride

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.