Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lutetium(III) chloride, ultra dry, 99.98% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used as an additive for chemical industry, single crystal scintillators, carbon arc rods, PET scan detectors, high refractive index glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Melting Point | 905°C (sublimation) |
Physical Form | Powder |
Quantity | 1 g |
Merck Index | 14,5615 |
Solubility Information | Soluble in water. |
Formula Weight | 281.33 |
Percent Purity | 99.98% |
Packaging | Ampule |
Sensitivity | Hygroscopic |
Density | 3.98 g/mL |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335-H500 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 233-240-1 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction