Alfa Aesar™ Lutetium(III) nitrate hydrate, REacton™, 99.99% (REO)

Manufacturer: Alfa Aesar™ 1125809

Quantity 10g

 View more versions of this product

Catalog No. AA1125809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100641-16-5
    Linear Formula Lu(NO3)3·xH2O
    Formula Weight 360.98 (Anhydrous)
    MDL Number MFCD00150421
    UN Number UN1477
    Chemical Name or Material Lutetium(III) nitrate hydrate
    Grade Reacton™

    Specifications

    Formula Weight 360.98 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula Lu(NO3)3·xH2O
    Health Hazard 1 Danger
    Physical Form Crystalline Aggregates
    Sensitivity Hygroscopic
    MDL Number MFCD00150421
    CAS 100641-16-5
    UN Number UN1477
    Color White
    Quantity 10g
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Lutetium(III) nitrate hydrate
    Assay 99.99%, (REO)
    Grade Reacton™

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit