Alfa Aesar™ Lutetium(III) oxide, 99.9% (REO)

Manufacturer: Alfa Aesar™ 3624406

Quantity 5g

 View more versions of this product

Catalog No. AA3624406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12032-20-1
    Formula Weight 397.94
    MDL Number MFCD00011100
    Merck Index 14,5615
    Chemical Name or Material Lutetium(III) oxide

    Specifications

    Formula Weight 397.94
    Physical Form Sintered Lump
    Merck Index 14,5615
    TSCA TSCA
    Odor Odorless
    Sensitivity Hygroscopic
    MDL Number MFCD00011100
    CAS 12032-20-1
    Melting Point 2487°C
    Quantity 5g
    Density 9.420g/mL
    Particle Size 3 to 12mm
    Chemical Name or Material Lutetium(III) oxide
    Assay 99.9%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit