Learn More
Lutetium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Description
Lutetium(III) oxide serves as a catalyst in a variety of reactions such as polymerization, hydrogenation and alkylation reactions. It is also used in ceramics and glass. In the production of laser crystals, lutetium(III) oxide finds application as an important starting material. Its luminescent properties, a field of interest for researchers and may find potential application in biological fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2487°C |
Physical Form | Powder |
Quantity | 25 g |
Merck Index | 14,5615 |
Solubility Information | Insoluble in water. |
Formula Weight | 397.94 |
Percent Purity | 99.9% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 9.420 |
Show More |
Safety and Handling
Hazard Category | H315-H320-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 234-764-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.