Alfa Aesar™ Lutetium(III) oxide, REacton™, 99.995% (metals basis), REM 50ppm

Manufacturer: Alfa Aesar™ 1091206

Quantity 5g

 View more versions of this product

Catalog No. AA1091206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12032-20-1
    Formula Weight 397.94
    MDL Number MFCD00011100
    Merck Index 14,5615
    Chemical Name or Material Lutetium(III) oxide, REM 50ppm
    Grade Reacton™

    Specifications

    Formula Weight 397.94
    Odor Odorless
    MDL Number MFCD00011100
    Melting Point 2487°C
    Density 9.420g/mL
    TSCA TSCA
    Physical Form Powder
    Sensitivity Hygroscopic
    CAS 12032-20-1
    Merck Index 14,5615
    Quantity 5g
    Chemical Name or Material Lutetium(III) oxide, REM 50ppm
    Assay 99.995%, (metals basis)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit