Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lutetium pieces, 99.9% (REO), Thermo Scientific™

Click to view available options
Quantity:
1 g
5 g
7439-94-3
Lu
174.967
MFCD00011098
OHSVLFRHMCKCQY-UHFFFAOYSA-N
lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
23929
CHEBI:33382
lutetium
[Lu]
7439-94-3
Lu
174.967
MFCD00011098
OHSVLFRHMCKCQY-UHFFFAOYSA-N
lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
23929
CHEBI:33382
lutetium
[Lu]
  • Packaged in air-stable glass bottle
Quantity 5 g
Odor Odorless
Packaging Glass bottle
Physical Form Pieces
Percent Purity 99.9%
Chemical Name or Material Lutetium pieces
Assay (REO)

P210-P240-P241-P280-P370+P378q

H228

EINECSNumber : 231-103-0

TSCA : Yes

storageNote1 :

  • Packaged in air-stable glass bottle

Recommended Storage : Ambient temperatures

RUO – Research Use Only