Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lutetium rod, 6.35mm (0.25 in.) dia., 99.9% (metals basis excluding Ta), Ta typically 2%

Manufacturer:  Alfa Aesar™ 43961DJ

Quantity 50mm

 View more versions of this product

Catalog No. AA43961DJ

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7439-94-3
MFCD00011098
Lutetium rod, ≈6.35mm (0.25 in.) dia.

Specifications

50mm
Odorless
Cast Rod
Lutetium rod, ≈6.35mm (0.25 in.) dia.
7439-94-3
Mylar under Argon
MFCD00011098
99.9%, (metals basis excluding Ta), Ta typically 2%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

storageNote1 :

  • Packaged in mylar under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit