Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LUZP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555883
Description
Immunogen sequence: RERHTSTSNI QVGLAELTSV SNHVSSPFEL SIHKHDITLQ LAEAERMADG PLKNRPETVV SRSSIIIKPS DPVERNSHAP PAETIRWKSH Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 86%.
This gene includes two alternatively spliced transcript variants, which encode different isoforms. The function of this gene has not been determined.
Specifications
LUZP1 | |
Polyclonal | |
Unconjugated | |
LUZP1 | |
2700072H04Rik; AI266952; Leucine zipper motif-containing protein; leucine zipper protein 1; Luzp; LUZP1; mFLJ00226; RP1-184J9.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7798 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86V48 | |
LUZP1 | |
Recombinant protein corresponding to Human LUZP1. Recombinant protein control fragment (Product #RP-94449). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction