Learn More
LUZP4 Mouse anti-Human, Clone: OTI3A10, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809088
Catalog No. 50-166-9261
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Export adapter involved in mRNA nuclear export in cancer cells. Binds and enhances the RNA-binding activity of the nuclear RNA export factor NXF1. Can restore mRNA export function in cells compromised by loss of mRNA export adapters.
Specifications
Specifications
LUZP4
Monoclonal
Unconjugated
LUZP4
Lyophilized
LUZP4
Full length human recombit protein of human LUZP4 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI3A10
PBS with 8% trehalose and no preservative; pH 7.3
Q9P127
CT-28, CT-8, CT28, HOM-TES-85
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
51213