Learn More
Abnova Human NCAPG2 Full-length ORF (AAH20560.1, 1 a.a. - 935 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00054892P01
Description
Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Both condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 (MIM 605576) and SMC4 (MIM 605575), but they contain different sets of non-SMC subunits. NCAPG2 is 1 of 3 non-SMC subunits that define condensin II (Ono et al., 2003 [PubMed 14532007]).[supplied by OMIM]
Sequence: MLLECFININYIKKEEGRRFLSCLFNWNINFIKMIHGTIKNQLQGLQKSLMVYIAEIYFRAWKKASGKILEAIENDCIQDFMFHGIHLPRRSPVHSKVREVLSYFHHQKKVRQGVEEMLYRLYKPILWRGLKARNSEVRSNAALLFVEAFPIRDPNLHAIEMDSEIQKQFEELYSLLEDPYPMVRSTGILGVCKITSKYWEMMPPTILIDLLKKVTGELAFDTSSADVRCSVFKCLPMILDNKLSHPLLEQLLPALRYSLHDNSEKVRVAFVDMLLKIKAVRAAKFWKICPMEHILVRLETDSRPVSRRLVSLIFNSFLPVNQPEEVWCERCVTLVQMNHAAARRFYQYAHEHTACTNIAKLIHVIRHCLNACIQRAVREPPEDEEEEDGREKENVTVLDKTLSVNDVACMAGLLEIIVILWKSIDRSMENNKEAKLYTINKFASVLPEYLKVFKDDRCKIPLFMLMSFMPASAVPPFSCGVISTLRSREEGAVDKSYCTLLDCLCSWGQVGHILELVDNWLPTEHAQAKSNTASKGRVQIHDTRPVKPELALVYIEYLLTHPKNRECLLSAPRKKLNHLLKALETSKADLESLLQTPGGKPRGFSEAAAPRAFGLHCRLSIHLQHKFCSEGKVYLSMLEDTGFWLESKILSFIQDQEEDYLKLHRVIYQQIIQTYLTVCKDVVMVGLGDHQFQMQLLQRSLGIMQTVKGFFYVSLLLDILKEITGSSLIQKTDSDEEVAMLLDTVQKVFQKMLECIARSFRKQPEEGLRLLYSVQRPLHEFITAVQSRHTDTPVHRGVLSTLIAGPVVEISHQLRKVSDVEELTPPEHLSDLPPFSRCLIGIIIKSSNVVRSFLDELKACVASNDIEGIVCLTAAVHIILVINAGKHKSSKVREVAATVHRKLKTFMEITLEEDSIERFLYESSSRTLGELLNSSpecifications
AAH20560.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
133.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAP-G2|FLJ20311|LUZP5|MTB|hCAP-G2 | |
NCAPG2 | |
Yes |
wheat germ expression system | |
Liquid | |
54892 | |
NCAPG2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLLECFININYIKKEEGRRFLSCLFNWNINFIKMIHGTIKNQLQGLQKSLMVYIAEIYFRAWKKASGKILEAIENDCIQDFMFHGIHLPRRSPVHSKVREVLSYFHHQKKVRQGVEEMLYRLYKPILWRGLKARNSEVRSNAALLFVEAFPIRDPNLHAIEMDSEIQKQFEELYSLLEDPYPMVRSTGILGVCKITSKYWEMMPPTILIDLLKKVTGELAFDTSSADVRCSVFKCLPMILDNKLSHPLLEQLLPALRYSLHDNSEKVRVAFVDMLLKIKAVRAAKFWKICPMEHILVRLETDSRPVSRRLVSLIFNSFLPVNQPEEVWCERCVTLVQMNHAAARRFYQYAHEHTACTNIAKLIHVIRHCLNACIQRAVREPPEDEEEEDGREKENVTVLDKTLSVNDVACMAGLLEIIVILWKSIDRSMENNKEAKLYTINKFASVLPEYLKVFKDDRCKIPLFMLMSFMPASAVPPFSCGVISTLRSREEGAVDKSYCTLLDCLCSWGQVGHILELVDNWLPTEHAQAKSNTASKGRVQIHDTRPVKPELALVYIEYLLTHPKNRECLLSAPRKKLNHLLKALETSKADLESLLQTPGGKPRGFSEAAAPRAFGLHCRLSIHLQHKFCSEGKVYLSMLEDTGFWLESKILSFIQDQEEDYLKLHRVIYQQIIQTYLTVCKDVVMVGLGDHQFQMQLLQRSLGIMQTVKGFFYVSLLLDILKEITGSSLIQKTDSDEEVAMLLDTVQKVFQKMLECIARSFRKQPEEGLRLLYSVQRPLHEFITAVQSRHTDTPVHRGVLSTLIAGPVVEISHQLRKVSDVEELTPPEHLSDLPPFSRCLIGIIIKSSNVVRSFLDELKACVASNDIEGIVCLTAAVHIILVINAGKHKSSKVREVAATVHRKLKTFMEITLEEDSIERFLYESSSRTLGELLNS | |
RUO | |
NCAPG2 | |
Wheat Germ (in vitro) | |
GST |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.