Learn More
Apricot DesignsSupplier Diversity Partner LW DEAD VOL RESERV 12CH PIPE  DFS Item
Manufacturer:  Apricot DesignsSupplier Diversity Partner RES18012W
12 individual channel polypopylene reagent reservoir (180mL max volume) 10 reservoirs/unit, 50 reservoirs/case. (low dead volume reservoir for 12 channel pipettor)
Catalog No. NC0583227