Learn More
LW Scientific Platelet Rotator
Integrated with heavy-duty motor holds up to 36 platelet bags
Manufacturer: LW Scientific RTLPLV112B1
Description
- Durable construction
- Whisper-quiet motor
- Variable speed (0-9rpm)
- Easy loading and unloading
- Stainless steel basket rotates 360°C
- Built-in tachometer allows users to monitor speed
- Widely used in blood banks
Baskets will not accommodate common size plateletpheresis bags
Specifications
0-9rpm | |
110V | |
One basket (12 bag capacity) |