Learn More
LW Scientific Platelet Rotator
Integrated with heavy-duty motor holds up to 36 platelet bags
Manufacturer:  LW Scientific RTLPLV336B1
 View more versions of this product
Catalog No. 22-500-105
Add to cart
Description
Description
  • Durable construction
  • Whisper-quiet motor
  • Variable speed (0-9rpm)
  • Easy loading and unloading
  • Stainless steel basket rotates 360°C
  • Built-in tachometer allows users to monitor speed
  • Widely used in blood banks
Baskets will not accommodate common size plateletpheresis bags
Specifications
Specifications
0-9rpm
110V
Three baskets (36 bag capacity)