Learn More
Abnova™ LXN 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00056925T01
Catalog No. 89-951-269
Add to cart
Description
Description
Specifications
Specifications
293T
NM_020169.2
pCMV-LXN full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LXN 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 24.53kDa
Transient overexpression cell lysate was tested with anti-LXN antibody (Mfr. No. H00056925-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human