Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LXR beta/NR1H2 Mouse anti-Human, Clone: LXRB/2731, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30717120UG
Description
LXR beta/NR1H2 Monoclonal specifically detects LXR beta/NR1H2 in Human samples. It is validated for ELISA, Protein Array.Specifications
LXR beta/NR1H2 | |
Monoclonal | |
0.2 mg/ml | |
ELISA, Protein Array | |
Liver X receptor beta, LXR-b, NER-I, NERliver X receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta | |
Recombinant full-length human LXR beta/NR1H2 protein | |
20 μg | |
Primary | |
Human | |
Purified |
ELISA, Peptide Array | |
LXRB/2731 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
7376 | |
Store at 4°C. | |
IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction