Learn More
Ly-108 Mouse anti-Mouse, BB700, Clone: 13G3, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 742272
Description
The 13G3 monoclonal antibody specifically binds to Lymphocyte antigen 108, Ly-108. Ly-108 is a member of the signaling lymphocytic activation molecule (SLAM) family of immune receptors. Ly-108 is a type 1 transmembrane glycoprotein adhesion receptor that is expressed by T cells, NKT cells, B cells, NK cells, macrophages, dendritic cells and neutrophils. Ly-108 plays multiple roles in innate and adaptive immunity including costimulation of NK cell cytotoxicity and T cell cytokine responses. Moreover, Ly-108 has been implicated in autoimmunity.
The antibody was conjugated to BD Horizon™ BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes. It is a polymer-based tandem dye developed exclusively by BD Biosciences. With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg, 695/40nm filter). This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.BD Horizon Brilliant™ Blue 700 (BB700) is a dye that was exclusively developed by BD Biosciences as brighter alternative to PerCP-Cy5.5. This dye also has less cross laser excitation off the 405 nm laser, resulting in less spillover into the violet channels compared to PerCP-Cy5.5. Due to similar excitation and emission properties, BD Horizon BB700 and PerCP-Cy5.5 cannot be used simultaneously.Specifications
Ly-108 | |
13G3 | |
Brilliant Blue 700 (BB700) | |
Slamf6, SLAM family member 6, KAL1, NTB-A, SF2000 | |
WT thymocytes | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.