Learn More
Azzota Corp LY-2090314 DFS Item
Manufacturer:  Azzota Corp C1201
LY-2090314 is a potent inhibitor of glycogen synthase kinase-3 (GSK-3) which plays an important role in many pathways, including initiation of protein synthesis, cell proliferation, cell differentiation, and apoptosis.
Catalog No. 50-161-3140