Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY 215840, Tocris Bioscience™

Catalog Number 152350
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
137328-52-0
C24H33N3O2
395.547
IMSDOBUYDTVEHN-ZLJPILQHSA-N
[8β(1S, 2R)]-N-(2-Hydroxycyclopentyl)-6-methyl-1-(1-methylethyl)-ergoline-8-carboxamide
122173083
(6aR,9R,10aS)-N-[(1S,2R)-2-hydroxycyclopentyl]-7-methyl-4-propan-2-yl-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline-9-carboxamide
CC(C)N1C=C2CC3C(CC(CN3C)C(=O)NC4CCCC4O)C5=C2C1=CC=C5
This item is not returnable. View return policy
137328-52-0
C24H33N3O2
395.547
IMSDOBUYDTVEHN-ZLJPILQHSA-N
[8β(1S, 2R)]-N-(2-Hydroxycyclopentyl)-6-methyl-1-(1-methylethyl)-ergoline-8-carboxamide
122173083
(6aR,9R,10aS)-N-[(1S,2R)-2-hydroxycyclopentyl]-7-methyl-4-propan-2-yl-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline-9-carboxamide
CC(C)N1C=C2CC3C(CC(CN3C)C(=O)NC4CCCC4O)C5=C2C1=CC=C5

5-HT2/5-HT7 antagonist

5-HT2/5-HT7 receptor antagonist. Reduces stimulatory effect of serotonin on aldosterone secretion; also reduces 5-HT-induced cAMP formation (pKB = 8.26).
Quantity 50 mg
Formula Weight 395.54
Percent Purity >98%
Chemical Name or Material LY 215840
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit