Learn More
Tocris Bioscience™ LY 2183240 Available on GSA/VA Contract for Federal Government customers only.
Novel, potent anandamide uptake inhibitor; inhibits FAAH
$119.00 - $255.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
24-521 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
2452/1
Each for $119.00
Add to cart
24-521-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
2452/10
Each for $255.00
Add to cart
Description
Description
Novel and highly potent blocker of anandamide uptake (IC50 = 270 pM). Inhibits fatty acid amide hydrolase (FAAH) activity (IC50 = 12.4 nM). Following i.p. administration in rats, increases brain anandamide concentration and exerts antinociceptive effects in formalin model of pain.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.