Learn More
Medchemexpress LLC Ly-2584702 tosylate salt | 1082949-68-5 | 99.5% | 617.62 | C28H27F4N7O3S | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12493A10MM 1ML
Encompass_Preferred
LY-2584702 tosylate salt is an ATP-competitive inhibitor of p70S6K supplied as a 10 mM solution in DMSO (1 mL). Reported activity includes an IC50 of 4 nM for p70S6K and 2 nM in an S6K1 enzyme assay; the compound is intended for use in biochemical and cellular studies of S6K/p70S6K signaling.
  • Provided as a 10 mM solution in DMSO, 1 mL.
  • High purity (99.5%).
  • Potent ATP-competitive inhibitor of p70S6K (IC50 4 nM).
  • Demonstrated activity in S6K1 enzyme assays (IC50 2 nM).
  • Suitable for biochemical kinase assays and cellular pathway studies.
  • Supplied as the tosylate salt for enhanced stability and handling.
Catalog No. 50-002-35198