Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LY 303511, Tocris Bioscience™
Click to view available options
Quantity:
5 mg
Description
Negative control compound for LY 294002 PI 3-kinase inhibitory activity. Blocks voltage-gated potassium (Kv) channels (IC50 = 64.6 μM) and inhibits IL-1β-stimulated NF-κB activation, attenuating MCP-1 expression. Antiproliferative. Also inhibits the BET bromodomain proteins BRD2, BRD3 and BRD4.
Active Analog also available.
Specifications
Quantity | 5 mg |
Formula Weight | 306.36 |
Percent Purity | >99% |
Chemical Name or Material | LY 303511 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction