Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY 334370 hydrochloride, Tocris Bioscience™

Catalog Number 307950
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
199673-74-0
C21H23ClFN3O
387.883
DGPDGAPZTPSHBL-UHFFFAOYSA-N
ly 334370 hydrochloride, 4-fluoro-n-3-1-methyl-4-piperidinyl-1h-indol-5-yl benzamidehydrochloride, 4-fluoro-n-3-1-methyl-4-piperidinyl-1h-indol-5-yl benzamide hydrochloride, 4-fluoro-n-3-1-methylpiperidin-4-yl-1h-indol-5-yl benzamide hydrochloride, ly-334370 hydrochloride hydrate, 4-fluoro-n-3-1-methylpiperidin-4-yl-1h-indol-5-yl benazmide hydrochloride, ly-334370 hydrochloride hydrate hplc , powder, 4-fluoro-n-3-1-methyl-4-piperidinyl-1h-indol-5-yl benzamide hydrochloride hydrate
19710026
4-fluoro-N-[3-(1-methylpiperidin-4-yl)-1H-indol-5-yl]benzamide;hydrochloride
CN1CCC(CC1)C2=CNC3=C2C=C(C=C3)NC(=O)C4=CC=C(C=C4)F.Cl
This item is not returnable. View return policy
199673-74-0
C21H23ClFN3O
387.883
DGPDGAPZTPSHBL-UHFFFAOYSA-N
ly 334370 hydrochloride, 4-fluoro-n-3-1-methyl-4-piperidinyl-1h-indol-5-yl benzamidehydrochloride, 4-fluoro-n-3-1-methyl-4-piperidinyl-1h-indol-5-yl benzamide hydrochloride, 4-fluoro-n-3-1-methylpiperidin-4-yl-1h-indol-5-yl benzamide hydrochloride, ly-334370 hydrochloride hydrate, 4-fluoro-n-3-1-methylpiperidin-4-yl-1h-indol-5-yl benazmide hydrochloride, ly-334370 hydrochloride hydrate hplc , powder, 4-fluoro-n-3-1-methyl-4-piperidinyl-1h-indol-5-yl benzamide hydrochloride hydrate
19710026
4-fluoro-N-[3-(1-methylpiperidin-4-yl)-1H-indol-5-yl]benzamide;hydrochloride
CN1CCC(CC1)C2=CNC3=C2C=C(C=C3)NC(=O)C4=CC=C(C=C4)F.Cl

Selective 5-HT1F agonist

Selective 5-HT1F receptor agonist (Ki values are 1.87, 16.4, > 100 (IC50), 176, 189, 281, 1280, 1530, 1550, > 3000 (IC50) and 3250 nM for 5-HT1F, 5-HT1A, 5-HT4, 5-HT1E, 5-HT1B, 5-HT1D, 5-HT2B, 5-HT2A, 5-HT7, 5-HT6 and 5-HT2C receptors respectively). Displays antimigraine effects.
Quantity 50 mg
Formula Weight 387.88
Percent Purity >99%
Chemical Name or Material LY 334370 hydrochloride
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit