Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LY 334370 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective 5-HT1F receptor agonist (Ki values are 1.87, 16.4, > 100 (IC50), 176, 189, 281, 1280, 1530, 1550, > 3000 (IC50) and 3250 nM for 5-HT1F, 5-HT1A, 5-HT4, 5-HT1E, 5-HT1B, 5-HT1D, 5-HT2B, 5-HT2A, 5-HT7, 5-HT6 and 5-HT2C receptors respectively). Displays antimigraine effects.
Specifications
Quantity | 10 mg |
Formula Weight | 387.88 |
Percent Purity | >99% |
Chemical Name or Material | LY 334370 hydrochloride |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction