Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LY 341495, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
Highly potent and selective group II metabotropic glutamate receptor antagonist (Ki/IC50 values are 2.3, 1.3, 173, 990, 6800, 8200 and 22000 nM for human mGlu2, mGlu3, mGlu8, mGlu7a, mGlu1a, mGlu5a and mGlu4a receptors respectively). Readily brain penetrant and active in vivo.
Disodium salt (Cat. No. 4062) also available.
Specifications
Quantity | 1 mg |
Formula Weight | 353.37 |
Percent Purity | >98% |
Chemical Name or Material | LY 341495 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction