Tocris™ LY 379268 disodium salt

Sodium salt of LY 379268 (Cat. No. 2453); selective group II mGlu receptor agonist

Manufacturer: Tocris™ 5064/50

 View more versions of this product

Catalog No. 506450R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors LY 379268 disodium salt
    Purity >98
    Molecular Weight 231.11
    Quantity 50mg
    Product Type LY 379268 disodium salt

    Sodium salt of LY 379268 (Cat. No. 2453), a highly selective group II mGlu receptor agonist.

    • Chemical Name: (1R,4R,5S,6R)-4-Amino-2-oxabicyclo[3.1.0]hexane-4,6-dicarboxylic acid disodium salt
    • Formula: C7H7NNa2O5
    • Solubility: Soluble to 100mM in water
    • Purity: >98
  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit