Learn More
Ly-49F Mouse anti-Mouse, BB700, Clone: HBF-719, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 745744
Catalog No. BDB745744
Add to cart
Description
Description
Ly-49F Monoclonal antibody specifically detects Ly-49F in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Ly-49F
HBF-719
BB700
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Mouse