Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-49H Mouse anti-Mouse, PE-Cyanine7, Clone: 3D10, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 25588680

 View more versions of this product

Catalog No. 50-112-3404


Add to cart

Description

Description

Description: The monoclonal antibody 3D10 recognizes Ly-49H, a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly-49H, like Ly-49D, lacks the characteristic ITIM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly-49H is expressed in a subset of NK cells but not NKT (NK1.1CD3+) cells. Natural killer cells can express several Ly-49 proteins. It has been shown that there is preferential staining of Ly-49H with Ly-49D (cat. 12-5783). Expression has been confirmed on C57BL/6 and NWNA but not Balb/c or DBA/2 mice. Other mice strains have not been tested. No crossreactivity to Ly-49A, C, D or G2 has been observed. Addition of monoclonal antibody 3D10 to cell cultures induces lysis of target cells. Applications Reported: This 3D10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3D10 antibody has been tested by fow cytometric analyis of mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered. Ly49H is a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly49H, like Ly49D, lacks the characteristic ITAM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly49H is expressed in a subset of NK cells but not NKT (NK1.1/CD3+) cells. Natural killer cells can express several Ly49 proteins.
Specifications

Specifications

Ly-49H
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q60682
Klra8
IgG1, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16639
Flow Cytometry
3D10
PE-Cyanine7
Klra8
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.