Learn More
Ly-49I Monoclonal Antibody (YLI-90), FITC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$150.65 - $336.00
Specifications
Antigen | Ly-49I |
---|---|
Clone | YLI-90 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Description: The YLI-90 monoclonal antibody reacts with the B6 mouse Ly-49I receptor. Ly49I, a member of the Ly49 family with inhibitory function, is a homodimer type II transmembrane protein and contains an Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM). YLI-90 binds to subpopulations of NK, NKT and T cell lineages in the C57Bl/6 strain. This antibody does not bind to cells from BALB/c, SJL or AKR/J mice. Applications Reported: YLI-90 has been reported for use in flow cytometric analysis. Applications Tested: The YLI-90 antibody has been tested by flow cytometric analysis of C57Bl/6 and BALB/c mouse splenocytes and can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.Specifications
Ly-49I | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
5E6; Killer cell lectin-like receptor 3; killer cell lectin-like receptor subfamily A, member 9; Klra3; Klra9; Ly-49; Ly49C; Ly-49c; Ly49I; Ly49I receptor; Ly-49I receptor; LY49I1; Lymphocyte antigen 49c; Nk2.1; T-cell surface glycoprotein Ly-49C | |
Klra9 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Klra9 |
YLI-90 | |
Flow Cytometry | |
FITC | |
Mouse | |
Mouse | |
0 | |
16640 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.