Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6A/E Rabbit anti-Mouse, Clone: JM53-11, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266939
Description
Ly-6A/E Monoclonal antibody specifically detects Ly-6A/E in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Ly-6A/E | |
JM53-11 | |
Unconjugated | |
Purified | |
Ly 6A/E, Ly6A.2, Ly6A/E, Ly6E.1, Lymphocyte antigen 6 complex locus A, Lymphocyte antigen 6 complex locus E, Retinoic acid induced gene E, RIGE, SCA1, SCA-1, SCA2, Stem cell antigen 1, Stem cell antigen 2, T cell activating protein, TAP, Thymic shared antigen 1, TSA1 | |
LY6A | |
Full length protein | |
Monoclonal | |
Protein A purified | |
Hematopoietic Stem Cell Markers, Stem Cell Markers | |
Mouse |
Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
110454 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title