Learn More
Ly-6A/E Rat anti-Mouse, BB515, Clone: D7, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 565397
Catalog No. BDB565397
Add to cart
Description
Description
Ly-6A/E Monoclonal antibody specifically detects Ly-6A/E in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
Ly-6A/E
D7
BB515
Purified
Rat
Rat LEW
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Ly-6A/E; Lymphocyte antigen 6A-2/6E-1; Ly-6A.2/Ly-6E.1; Sca-1; TAP
IL-2-dependent mouse T-cell line CTL-L
50μg
Cell Biology
Primary
Murine
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.