Learn More
Ly-6A/E Rat anti-Mouse, BUV395, Clone: D7, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566216
Catalog No. BDB566216
Add to cart
Description
Description
Ly-6A/E Monoclonal antibody specifically detects Ly-6A/E in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
Ly-6A/E
D7
Brilliant Ultraviolet 395
Purified
Rat
Rat LEW, also known as Lewis IgG2a, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions, and unconjugated antibody and free BD Horizon™ BUV395 were removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Ly-6A/E; Lymphocyte antigen 6A-2/6E-1; Ly-6A.2/Ly-6E.1; Sca-1; TAP
IL-2-dependent mouse T-cell line CTL-L
25μg
Cell Biology
Primary
Murine
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.